Learn More
JDM LAB SOLUTIONS LLC Oxford BenchMate 10 Position Magnetic Hotplate Stirrer, Speed range - 300-1500 rpm, includes 1 Stir Bar. 110/230 V, 50/60Hz.
Supplier:  JDM LAB SOLUTIONS LLC MHS10
Encompass
Oxford BenchMate 10 Position Magnetic Hotplate Stirrer, Speed range - 300-1500 rpm, includes 1 Stir Bar. 110/230 V, 50/60Hz.
Catalog No. NC3934358