Learn More
Cell Biolabs Inc OxiSelect™ In Vitro ROS/RNS Assay Kit (Green Fluorescence), 96 assays
Supplier:  Cell Biolabs Inc STA347
Encompass_Preferred
The OxiSelect™ In Vitro ROS/RNS Assay Kit is an assay for measuring the total free radical presence of a sample. The assay employs a proprietary quenched fluorogenic probe, DCFH-DiOxyQ), which is based on similar chemistry to the popular DCFH-DA. The DCFH-DiOxyQ probe is first primed with a quench removal reagent, and subsequently stabilized in the highly reactive DCFH form. In this reactive state, ROS and RNS species can react with DCFH, which is rapidly oxidized to the highly fluorescent 2’, 7’-dichlorodihydrofluorescein (DCF) . The DCFH-DiOxyQ probe can react with hydrogen peroxide, peroxyl radical, nitric oxide, and peroxynitrite anion. These free radical molecules are representative of both ROS and RNS, thus allowing for measurement of the total free radical population within a sample. The kit has a detection sensitivity limit of 40 nM for H2O2. Each kit provides sufficient reagents to perform up to 96 assays, including standard curve and unknown samples.
Catalog No. NC0006519
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.