Learn More
Selleck Chemical LLC Oxibendazole S1851-50mg
SDP
Supplier:  Selleck Chemical LLC S185150MG
Encompass_Preferred
Oxibendazole is a benzimidazole drug that interferes with metabolic pathways used to protect against roundworms strongyles threadworms pinworms and lungworm infestations in horses and some domestic pets
Catalog No. 50-311-0818