Learn More
Apexbio Technology LLC Oxibendazole 20559-55-1 200mg
SDP
Supplier:  Apexbio Technology LLC B1999200
Encompass_Preferred
Oxibendazole (CAS 20559-55-1) is a small-molecule inhibitor targeting microtubule polymerization in nematodes It is designed to disrupt cytoskeletal assembly thereby impairing parasite survival Oxibendazole exerts its biological activity primarily through inhibition of microtubule polymerization In in vitro studies oxibendazole demonstrates induction of apoptotic signaling pathways and inhibits proliferation in mammalian tumor cell lines with reported IC50 values for its antiproliferative effect typically ranging from low to mid micromolar levels Based on these pharmacological properties oxibendazole holds research potential in oncology and inflammatory disease models
Catalog No. 50-000-93110