Learn More
Medchemexpress LLC Oxibendazole | 20559-55-1 | 99.6% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB029910MM/1ML
Encompass_Preferred
Oxibendazole is an effective benzimidazole anthelmintic active against nematode infections. It has been shown to induce apoptosis and exhibit both anti-cancer and anti-inflammation activities. This product is intended for research use only.
  • Effective anthelmintic
  • Active against nematode infections
  • Induces apoptosis
  • Exhibits anti-cancer activity
  • Exhibits anti-inflammation activity
Catalog No. 50-004-06035