Learn More
Medchemexpress LLC Oxibendazole | 20559-55-1 | MFCD00133728 | 99.6% | 249.27 | C12H15N3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB029910MG
Encompass_Preferred
Oxibendazole is a benzimidazole anthelmintic research compound (CAS 20559-55-1) with reported apoptosis-inducing, anti-cancer, and anti-inflammatory activities. It is supplied for laboratory research in solid form or as DMSO solutions, with a reported purity of approximately 99.6% and a molecular weight of 249.27 g/mol.
  • Effective benzimidazole anthelmintic against nematode infections.
  • Induces apoptosis in preclinical studies, indicating potential anti-cancer activity.
  • Available as solid or ready-to-use DMSO solution for experimental flexibility.
  • High reported purity suitable for research applications.
  • Well-defined chemical identity: molecular weight 249.27 g/mol, CAS 20559-55-1.
Catalog No. 50-002-98893