Learn More
Medchemexpress LLC Oxibendazole | 20559-55-1 | MFCD00133728 | 99.6% | 50 MG

Supplier: Medchemexpress LLC HYB029950MG
Oxibendazole is an effective benzimidazole anthelmintic against nematode infections. It induces apoptosis and has anti-cancer and anti-inflammation activities. For research use only; not sold to patients.
- Molecular weight: 249.27
- Formula: C12H15N3O3
- Appearance: Solid
- Color: White to yellow
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 6 months, -20°C for 1 month
- Solubility (In Vitro): DMSO: 5 mg/mL (20.06 mM; ultrasonic and warming and heat to 60°C; Hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.