Learn More
Medchemexpress LLC Oxiconazole nitrate | 64211-46-7 | 98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB132450MG
Encompass_Preferred
Oxiconazole nitrate is a broad-spectrum antifungal agent effective against various fungi, including Candida, Aspergillus, and Trichophyton. It also acts as a potent activator of CYP3A4 transactivation, a process that can be competitively inhibited by Rifampicin. Furthermore, oxiconazole nitrate has demonstrated an inhibitory effect against colorectal cancer by inducing autophagy arrest mediated by peroxiredoxin-2.
  • Broad-spectrum antifungal activity.
  • Inhibits the growth of Candida, Aspergillus, and Trichophyton.
  • Activator of CYP3A4 transactivation.
  • Exhibits inhibitory effects against colorectal cancer.
Catalog No. 50-187-4500