Learn More
Medchemexpress LLC Oxiconazole nitrate | 64211-46-7 | 98.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB13245MG
Encompass_Preferred
Oxiconazole nitrate is a broad-spectrum antifungal agent. It is known to inhibit the growth of various fungi, including Candida, Aspergillus, and Trichophyton. This compound also acts as an efficacious activator of CYP3A4 transactivation and demonstrates an inhibitory effect against colorectal cancer (CRC) through peroxiredoxin-2 (PRDX2)-mediated autophagy arrest.
  • Inhibits growth of Candida, Aspergillus, and Trichophyton
  • Activates CYP3A4 transactivation
  • Shows inhibitory effect against colorectal cancer (CRC)
  • Mediates autophagy arrest via peroxiredoxin-2 (PRDX2)
Catalog No. 50-004-12482