Learn More
Medchemexpress LLC Oxiconazole nitrate (Ro 13-8996) | 64211-46-7 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB132410MM/1ML
Encompass_Preferred
Oxiconazole nitrate (Ro 13-8996) is a broad-spectrum anti-fungal agent effective against *Candida*, *Aspergillus*, and *Trichophyton*. It also acts as a highly effective activator of CYP3A4 transactivation and shows an inhibitory effect against colorectal cancer (CRC) by causing peroxiredoxin-2 (PRDX2)-mediated autophagy arrest.
  • Broad-spectrum anti-fungal agent
  • Effective against *Candida*, *Aspergillus*, and *Trichophyton*
  • Activates CYP3A4 transactivation
  • Inhibitory effect against colorectal cancer (CRC) via PRDX2-mediated autophagy arrest
  • Inhibits CRC cell growth in vitro and in vivo
Catalog No. 50-004-12080