Learn More
Medchemexpress LLC Oxidopamine (hydrobromide) | 636-00-0 | 99.4% | 50 MG
SDP
Catalog No. 5000413809
Encompass_Preferred
Oxidopamine (6-OHDA) hydrobromide is a neurotoxin that selectively destroys dopaminergic neurons and acts as an antagonist of the neurotransmitter dopamine. It promotes COX-2 activation, leading to PGE2 synthesis and pro-inflammatory cytokine IL-1β secretion. This compound is used in research for Parkinson's disease (PD), attention-deficit hyperactivity disorder (ADHD), and Lesch-Nyhan syndrome.
  • Antagonist of the neurotransmitter dopamine.
  • Selectively destroys dopaminergic neurons.
  • Promotes COX-2 activation, leading to PGE2 synthesis and pro-inflammatory cytokine IL-1β secretion.
  • Used for research of Parkinson's disease (PD), attention-deficit hyperactivity disorder (ADHD), and Lesch-Nyhan syndrome.
  • Can induce Parkinson's disease models.
Catalog No. 50-004-13809 Supplier Medchemexpress LLC Supplier No. HYB1081A50MG
Oxidopamine (6-OHDA) hydrobromide is a neurotoxin that selectively destroys dopaminergic neurons and acts as an antagonist of the neurotransmitter dopamine. It promotes COX-2 activation, leading to PGE2 synthesis and pro-inflammatory cytokine IL-1β secretion. This compound is used in research for Parkinson's disease (PD), attention-deficit hyperactivity disorder (ADHD), and Lesch-Nyhan syndrome.
  • Antagonist of the neurotransmitter dopamine.
  • Selectively destroys dopaminergic neurons.
  • Promotes COX-2 activation, leading to PGE2 synthesis and pro-inflammatory cytokine IL-1β secretion.
  • Used for research of Parkinson's disease (PD), attention-deficit hyperactivity disorder (ADHD), and Lesch-Nyhan syndrome.
  • Can induce Parkinson's disease models.