Learn More
Medchemexpress LLC Oxolinic acid | 14698-29-4 | 98.97% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1002500MG
Encompass_Preferred
Oxolinic acid is an antibiotic effective against both Gram-negative and Gram-positive bacteria. It is used in research for acute and chronic urinary tract infections. It functions as a DNA/RNA synthesis inhibitor and also acts as a dopamine uptake inhibitor, stimulating locomotor effects in mice.
  • Antibiotic active against Gram-negative and Gram-positive bacteria.
  • Used for research in acute and chronic urinary tract infections.
  • Functions as a DNA/RNA synthesis inhibitor.
  • Acts as a dopamine uptake inhibitor.
  • Stimulates locomotor effects in mice.
Catalog No. 50-004-07501