Learn More
Research Products International Corp Oxolinic Acid 500 G
SDP
Supplier:  Research Products International Corp O21450500.0
Encompass_Preferred
Oxolinic acid is an antibiotic in the quinolone class.
It works by inhibiting the bacterial DNA gyrase enzyme, which is essential for DNA replication and repair in bacteria. This disruption of DNA processes ultimately leads to the death of the bacteria.
Catalog No. 50-000-52434