Learn More
Medchemexpress LLC Oxolinic acid-d5 | 1189890-98-9 | 98.0% | 500 UG
SDP
Supplier:  Medchemexpress LLC HYB1002S500UG
Encompass_Preferred
Oxolinic acid-d5 is a deuterium labeled Oxolinic acid, an antibiotic effective against both Gram-negative and Gram-positive bacteria. It is used in the research of acute and chronic urinary tract infections.
  • Acts as a DNA/RNA synthesis inhibitor.
  • Functions as a dopamine uptake inhibitor.
  • Stimulates locomotor effects in mice.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07538