Learn More
Medchemexpress LLC Oxybenzone | 131-57-7 | 99.9% | C14H12O3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYA0067500MG
Encompass_Preferred
Oxybenzone (Benzophenone 3) is a widely used UV filter found in sun tans and skin protectants. It functions as an endocrine-disrupting chemical (EDC) and is capable of crossing both placental and blood-brain barriers. Research indicates that this compound impairs autophagy, alters epigenetic status, and disrupts retinoid X receptor signaling in apoptotic neuronal cells.
  • Acts as a UV filter in sunscreens and skin protectants
  • Functions as an endocrine disrupting chemical (EDC)
  • Capable of crossing both placental and blood-brain barriers
  • Impairs autophagy
  • Alters epigenetic status
  • Disrupts retinoid X receptor signaling in apoptotic neuronal cells
  • For research use only
Catalog No. 50-004-32080