Learn More
Apexbio Technology LLC Oxybutynin chloride 1508-65-2 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B11345.1
Encompass_Preferred
Oxybutynin chloride (CAS 1508-65-2) is a synthetic antimuscarinic agent that functions primarily by inhibiting the action of acetylcholine on muscarinic receptors in smooth muscle particularly within the bladder This antagonism results in reduced involuntary detrusor muscle contractions thereby modulating urinary urgency and frequency In biomedical research oxybutynin chloride is employed to investigate cholinergic pathways involved in bladder function neurogenic bladder disorders and the pharmacological modulation of muscarinic signaling in urinary physiology
Catalog No. 50-000-88359