Learn More
Medchemexpress LLC Oxybutynin chloride | 1508-65-2 | 98.5% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0267A10MM/1ML
Encompass_Preferred
Oxybutynin chloride is an orally active and competitive mAChR antagonist that inhibits vascular Kv channels. It reduces muscle spasm in the bladder and urinary tract and is useful in the study of overactive bladder syndrome (OAB). The compound also acts as a click chemistry reagent, containing an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.
  • Oral active and competitive mAChR antagonist
  • Has Kis of 14.3 nM and 5.55 nM for specific [3H]NMS binding in mouse bladder and cerebral cortex
  • Inhibits vascular Kv channels independently of anticholinergic effects, with an IC50 of 11.51 μM
  • Reduces muscle spasm in the bladder and urinary tract
  • Can be used in studies of overactive bladder syndrome (OAB)
  • Click chemistry reagent containing an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-004-05634