Learn More
Medchemexpress LLC Oxyclozanide | 2277-92-1 | 98.7% | 401.46 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY17594200MG
Encompass_Preferred
Oxyclozanide is an orally active salicylanilide anthelmintic agent that primarily functions by uncoupling oxidative phosphorylation in flukes. It also exhibits good anti-adenovirus, anti-biofilm, antifungal, and antibacterial activities.
  • Orally active salicylanilide anthelmintic agent.
  • Mainly acts by uncoupling oxidative phosphorylation in flukes.
  • Shows good anti-adenovirus, anti-biofilm, antifungal, and antibacterial activity.
  • Effective against meticillin-sensitive and meticillin-resistant S. pseudintermedius isolates.
  • Inhibits methicillin-resistant S. aureus (MRSA).
  • Demonstrates antiparasitic effectiveness in sheep and cattle.
Catalog No. 50-003-87133