Learn More
Medchemexpress LLC Oxyfedrine hydrochloride | 16777-42-7 | MFCD01695148 | 99.9% | 349.85 g/mol | C19H24ClNO3 | 1 ML

Supplier: Medchemexpress LLC HY10830010MM 1ML
Oxyfedrine hydrochloride is a β-adrenoreceptor agonist supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for research applications. The accurately prepared solution simplifies assay setup and reduces measurement variability. Storage and handling recommendations are provided to maintain product stability.
- Prepared 10 mM solution in DMSO, 1 mL.
- High purity (99.9% by HPLC).
- Suitable for in vitro assays and compound screening.
- Includes storage and stability guidance for solvent and solid forms.
- Molecular formula C19H24ClNO3; molecular weight 349.85 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.