Learn More
Med Vet International OxyHood, medium, J0652B
Supplier: Med Vet International JOR652B
The OxyHood was developed to deliver oxygen in aless restrictive and stressful environment. The Oxy Hooduses the conventional cone shape of an Elizabethan collarand adds a clear plastic "lid" over the end. Numerous vent holes throughout the cone allows the excess gas to vent. Oxygen is provided to the Oxy Hood via a nasal cannula attached to the hood. The oxygen source uses a bubble humidifier to prevent drying of the respiratory tract.Features: Collar is sewn to nylon fabric with a drawstring, plus velcro attachment allows for proper neck sizing Lid, or face of unit is attached with zipper Clear plastic material is used throughout for better observation and for the animal to see clearly Four sets of die cut holes allow ventilation of carbon dioxide and to maintain the oxygen level at more than 60% Very small animals can be placed totally in the larger Oxy Hood like an oxygen tent Complete with humidifier, oxygen nasal cannula, and oxygen tubing