Learn More
CPC Scientific H-His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-Lys-Arg-Asn-Arg-Asn-Asn-Ile-Ala-OH 0.5MG
SDP
Supplier:  CPC Scientific GLUC015A
Encompass
Oxyntomodulin is a potent inhibitor of gastric acid secretion and pancreatic enzyme secretion when infused iv. It was also proposed that intracerebroventricularly and into the hypothalamic paraventricular nucleus injected oxyntomodulin inhibits food intake in fasted and nonfasted animals potently and in a sustained manner.SEQUENCE: H-His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-Lys-Arg-Asn-Arg-Asn-Asn-Ile-Ala-OHONE-LETTER SEQUENCE: HSQGTFTSDYSKYLDSRRAQDFVQWLMNTKRNRNNIAMOLECULAR FORMULA: C192H295N61O60S1MOLECULAR WEIGHT: 4449.90STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [159002-68-3]SYNONYMS: Glucagon-37 (human, mouse, rat), Preproglucagon (53-89) (human, mouse, rat), OXM (human, mouse, rat), Proglucagon (33-69) (human, mouse, rat)RESEARCH AREA: DiabetesREFERENCES:C.L.Dakin et al., Endocrinology, 142, 4244 (2001)SKU(s): GLUC-015A, GLUC-015B
Catalog No. 50-210-9384