Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxypeucedanin hydrate | 2643-85-8 | 98.0% | 304.29 | C16H16O6 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN262210MG

Encompass_Preferred

Oxypeucedanin hydrate is a naturally occurring furanocoumarin isolated from D. anethifolia, supplied for laboratory research. It is provided as a small, high-purity powder suitable for analytical and biological studies.

  • Natural furanocoumarin isolated from D. anethifolia.
  • High purity (98.0%) suitable for analytical and biological assays.
  • Small 10 mg pack size for screening and method development.
  • Documented mild cytotoxicity in fibroblasts and parental lymphoma cells.
  • Stable as powder when stored at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Solvent aliquots recommended to be stored at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-89168


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.