Learn More
Medchemexpress LLC Oxyphenbutazone monohydrate | 7081-38-1 | 98.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB13555MG
Encompass_Preferred
Oxyphenbutazone monohydrate is a Phenylbutazone metabolite that exhibits anti-inflammatory effects. This orally active, non-selective COX inhibitor can selectively kill non-replicating Mycobacterium tuberculosis. It also enhances the anticancer efficiency of Methotrexate in Hep3B cells and exhibits reparative effects in hepatocytes.
  • Phenylbutazone metabolite with anti-inflammatory properties
  • Orally active non-selective COX inhibitor
  • Selectively kills non-replicating Mycobacterium tuberculosis
  • Enhances anticancer efficiency in Hep3B cells
  • Exhibits reparative effects in hepatocytes
Catalog No. 50-004-12127