Learn More
Oxyrase Oxyrase Phenylethyl Alcohol (PEA) Blood Agar OxyPlate
SDP
Supplier:  Oxyrase OPEABA
Encompass
Microbiology Media, Plated Media, Selective Media, Oxyrase, PEA OxyPlate, OxyPlate patented design for primary set up and subbing,, Manufactured and Packaged PRAS, Establishes anaerobiosis with no additional consumables, regenerates and maintains anaerobic conditions repeatedly, anaeobic specimen primary set up and subbing, Consists of 10 individually packaged plates, 3 month shelf life.
Catalog No. NC3813074