Learn More
Apexbio Technology LLC Oxysophocarpine 26904-64-3 20mg
SDP
Supplier:  Apexbio Technology LLC N128020
Encompass_Preferred
Oxysophocarpine is a quinolizidine alkaloid derived from the roots of Sophora flavescens It has been explored in biomedical research for its pharmacological effects particularly in anti-inflammatory analgesic and antitumor activities Oxysophocarpine exerts its biological activity through modulation of cellular signaling pathways regulation of inflammatory mediators and interference with specific receptor interactions Based on these pharmacological properties Oxysophocarpine holds research potential in anti-inflammatory analgesic and antitumor applications
Catalog No. 50-000-97094