Learn More
Cayman Chemical OxytetracyclIn 50g
SDP
Supplier:  Cayman Chemical 1807650 G
Encompass_Preferred
A broad-spectrum tetracycline antibiotic that inhibits protein synthesis in both Gram-positive and Gram-negative bacteria
Catalog No. 50-296-6272