Learn More
U.S. Pharmacopeia Oxytetracycline, 6153-64-6, 200mg
Supplier:  U.S. Pharmacopeia 1491004
Encompass
Molecular formula C22-H24-N2-O9, Molecular weight 460.44 g/mol, Melting Point 354.2 - 364.1 °F (179 - 184.5 °C) (decomposes) / 364.1 °F (184.5 °C), Synonyms: Hydroxytetracycline
Catalog No. NC9911498