Learn More
Medchemexpress LLC Oxytetracycline | 79-57-2 | 99.2% | 460.43 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHYB0275
Encompass_Preferred
Oxytetracycline is an antibiotic belonging to the tetracycline class. It potently inhibits Gram-negative and Gram-positive bacteria. This compound acts as a protein synthesis inhibitor, preventing the binding of aminoacyl-tRNA to the ribosomal RNA complex. It also exhibits anti-HSV-1 activity. This product is for research use only.
  • Potently inhibits gram-negative and gram-positive bacteria
  • Functions as a protein synthesis inhibitor
  • Prevents aminoacyl-tRNA binding to the ribosomal RNA complex
  • Exhibits anti-HSV-1 activity
  • Microbial metabolite and aromatic polyketide
  • Available as a solid
Catalog No. 50-003-64700