Learn More
Selleck Chemical LLC Oxytetracycline hydrochloride S5199-25mg
SDP
Supplier:  Selleck Chemical LLC S519925MG
Encompass_Preferred
Oxytetracycline (OTC Oxycycin Stevacin Terramycin) hydrochloride is a prescription antibiotic interfering with the ability of bacteria to produce essential proteins
Catalog No. 50-311-2350