Learn More
Medchemexpress LLC Oxytetracycline hydrochloride | 2058-46-0 | 98.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0275AR100MG
Encompass_Preferred
Oxytetracycline hydrochloride is an antibiotic belonging to the tetracycline class. It potently inhibits Gram-negative and Gram-positive bacteria. It acts as a protein synthesis inhibitor, preventing the binding of aminoacyl-tRNA to the m-ribosomal RNA complex. Additionally, it exhibits anti-HSV-1 activity.
  • Potently inhibits Gram-negative and Gram-positive bacteria.
  • Functions as a protein synthesis inhibitor.
  • Prevents the binding of aminoacyl-tRNA to the m-ribosomal RNA complex.
  • Exhibits anti-HSV-1 activity.
  • Classified as a tetracycline antibiotic.
Catalog No. 50-004-09001