Learn More
Apexbio Technology LLC Oxythiamine 136-16-3 50mg
SDP
Supplier:  Apexbio Technology LLC A473750
Encompass_Preferred
Oxythiamine (CAS 136-16-3) is a small molecule that functions as a thiamine antagonist by inhibiting transketolase (TK) activity a key enzyme in the non-oxidative branch of the pentose phosphate pathway By blocking TK oxythiamine disrupts ribose biosynthesis leading to impaired nucleotide production and interference with cellular metabolic processes Experimental studies indicate that oxythiamine modulates apoptotic signaling pathways and suppresses the proliferation of cancer cells This compound serves as a valuable tool in cancer metabolism research for investigating the effects of thiamine-dependent enzyme inhibition
Catalog No. 50-000-94866