Learn More
Bachem Oxytocin Acetate
SDP
Supplier:  Bachem 4016373.0005
Encompass_Preferred
Oxytocin Acetate, H-Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Leu-Gly-NH2, 50-56-6, C43H66N12O12S2, Mr 1007.20, 5mg. OXT. Oxytocin (OXT) is used to assist during labor to stimulate contractions of the uterus and mammary glands for milk letdown. Endogenous oxytocin also plays an important role in social behavior.
Catalog No. NC9932627