Learn More
Sigma Aldrich Fine Chemicals Biosciences Oxytocin acetate salt hydr5MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences O63795MG
Encompass
Oxytocin (OXT) is a potent natriuretic hormone encoded by the gene mapped to human chromosome 20p13. It is synthesized along with its carrier protein neurophysin I from its inactive precursor prepro-OXT. OXT gene consists of three exons and two introns where first exon codes for hormone OXT while other exon codes for neurophysin I.
Catalog No. 50-290-2199