Learn More
Medchemexpress LLC Oxytocin free acid (9-Deamidooxytocin) | 4248-64-0 | >98% | 1007.19 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP3216B100MG
Encompass_Preferred
Oxytocin free acid (9-Deamidooxytocin) is an analog of oxytocin where the glycinamide residue at position 9 has been replaced by a glycine residue. This pleiotropic peptide hormone has broad implications for general health, adaptation, development, reproduction, and social behavior.
  • Analog of oxytocin
  • Pleiotropic peptide hormone
  • Impacts general health and adaptation
  • Plays a role in development and reproduction
  • Influences social behavior
Catalog No. 50-003-98728