Learn More
Cayman Chemical OxytocInacetate 10mg
SDP
Supplier:  Cayman Chemical 1179910 MG
Encompass_Preferred
A nonapeptide hormone primarily synthesized in the hypothalamus with roles in uterine contraction and lactation, social memory and attachment, sexual and maternal behavior, aggression, learning, anxiety, feeding, and pain perception
Catalog No. 50-295-3280