Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ozagrel hydrochloride | 78712-43-3 | MFCD06795644 | >98.0% | 10 MG
SDP

Supplier:  Medchemexpress LLC HYB0428B10MG

Encompass_Preferred

Ozagrel hydrochloride is the hydrochloride salt of ozagrel, a selective thromboxane A2 (TXA2) synthase inhibitor used in preclinical research to study platelet aggregation, ischemic stroke, asthma, and thromboembolic disease.

  • Selective thromboxane A2 (TXA2) synthase inhibitor with reported IC50 ~11 nM.
  • Reduces thromboxane A2 production, leading to anti-platelet aggregation and vasodilation.
  • Hydrochloride salt supplied as a powder suitable for in vitro and in vivo research.
  • Typical molecular weight 264.71 g/mol and formula C13H13ClN2O2.
  • High purity commonly reported as ≥98% (verify with vendor for specific lot).
  • Store powder at -20°C for long-term stability; in solution, store at -80°C.

Catalog No. 50-002-98237


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.