Learn More
Medchemexpress LLC Ozagrel methyl ester | 866157-50-8 | 242.27 | 1 G
Supplier: Medchemexpress LLC HYW0046451G
Ozagrel methyl ester is a compound identified as a drug impurity, intended exclusively for research purposes. It is supplied as a solid and has a molecular weight of 242.27.
- For research use only
- Molecular formula: C14H14N2O2
- Appears as a solid
- Stable for 3 years at room temperature storage
- In solvent, stable for 2 years at -80°C or 1 year at -20°C