Learn More
Selleck Chemical LLC Ozagrel sodium S5252-25mg
SDP
Supplier:  Selleck Chemical LLC S525225MG
Encompass_Preferred
Ozagrel sodium (KCT-0809 Cataclot Xanbo) is a thromboxane A2 synthase (CYP5) inhibitor with neuroprotective properties
Catalog No. 50-311-2244