Learn More
Medchemexpress LLC Ozanimod | 1306760-87-1 | 98.0% | 500 MG
SDP
Supplier:  Medchemexpress LLC HY12288500MG
Encompass_Preferred
Ozanimod (RPC-1063) is a small-molecule sphingosine 1-phosphate (S1P) receptor modulator used as a research reference standard for studies of immune modulation and multiple sclerosis. It selectively binds S1P1 and S1P5 receptors and is supplied for analytical and in vitro research (CAS 1306760-87-1).
  • Selective S1P1 and S1P5 receptor modulation
  • Low-nanomolar in vitro potency
  • Available as a solid reference standard in multiple sizes, including 500 mg
  • Powder stable at -20°C for long-term storage
  • 98% purity standard suitable for analytical use
Catalog No. 50-002-30200