Learn More
Selleck Chemical LLC Ozanimod hydrochloride E4956-5MG
SDP
Supplier:  Selleck Chemical LLC E49565MG
Encompass_Preferred
Ozanimod hydrochloride(Zeposia RPC-1063 HCL) is a potent sphingosine 1-phosphate (S1P)receptor modulator and a selective agonist of S1P receptor subtypes 1 (S1P1) and 5 (S1P5) with EC50 s of 1 03 nM and 8 6 nM respectively It can be used in the trearment of relapsing multiple sclerosis (MS)
Catalog No. 50-000-43322