Learn More
SKC Inc Ozone Test Strips, Tube of 50 Strips
SDP
Supplier:  SKC Inc 526300
Encompass
Ozone Test Strips display results in four colors, each representing a level of ozone concentration from < 0.045 ppm to > 0.105 ppm, tube of 50 sticks. Quick indication of ambient ozone levels; Color results display in 10 minutes and Use for indoor or outdoor air. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC9608216