Learn More
TARGETMOL CHEMICALS INC P-1075 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T164205MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. P-1075 is a potent sulfonylurea receptor 2-associated ATP-sensitive potassium channels (SUR2-KIR6) activator (EC50 = 45 nM). P-1075 opens opens mitoKATP channels and may be cardioprotective. Purity 99.35%
Catalog No. 50-287-8752