Learn More
Medchemexpress LLC P-1075 | 60559-98-0 | 99.5% | 231.30 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10857310MM/1ML
Encompass_Preferred
P-1075 is a potent activator of sulfonylurea receptor 2-associated ATP-sensitive potassium channels (SUR2-KIR6), demonstrating an EC50 value of 45 nM for SUR2B-KIR6 channel activation. It also opens mitochondrial K(ATP) channels and generates reactive oxygen species, leading to cardioprotection in rabbit hearts.
  • Potent activator of sulfonylurea receptor 2-associated ATP-sensitive potassium channels (SUR2-KIR6)
  • EC50 value of 45 nM for SUR2B-KIR6 channel activation
  • Opens mitochondrial K(ATP) channels
  • Generates reactive oxygen species
  • Results in cardioprotection of rabbit hearts
  • Solid, white to off-white appearance
Catalog No. 50-003-46155