Learn More
Medchemexpress LLC P 22077 | 1247819-59-5 | 98.4% | 315.32 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1386550MG
Encompass_Preferred
P 22077 is a cell-permeable ubiquitin-specific protease 7 (USP7) inhibitor with an EC50 of 8.01 μM. It also inhibits USP47 with an EC50 of 8.74 μM.
  • USP7 inhibitor (EC50: 8.01 μM)
  • USP47 inhibitor (EC50: 8.74 μM)
  • Cell-permeable
  • Inhibits a smaller subset of DUBs at 15-45 μM
  • Causes DUBs inhibition in HEK293T cells at 25 μM
  • Greatly reduces cell viability of neuroblastoma (NB) cells
  • Increases p53 activity and induces apoptosis in p53 wild-type and HDM2-expressing NB cells at 10 μM
  • Enhances cytotoxic effect of Dox and VP-16 on NB cells, and enhances Dox- and VP-16-induced p53-mediated apoptosis at 5 μM
  • Shows potent antitumor activities in xenograft mouse models bearing IMR-32-derived tumors
  • Exhibits antitumor effects in mice bearing SH-SY5Y-derived tumors and NGP-derived tumors
Catalog No. 50-003-48958