Learn More
Medchemexpress LLC P 22077 | 1247819-59-5 | 98.42% | 315.32 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY13865
Encompass_Preferred
P 22077 is a cell-permeable inhibitor targeting ubiquitin-specific protease 7 (USP7) with an EC50 of 8.01 μM, and USP47 with an EC50 of 8.74 μM. This compound demonstrates antiproliferative activity against human HCT116 cells and induces p53-mediated apoptosis in neuroblastoma cells, highlighting its potential for anticancer research.
  • Cell-permeable USP7 and USP47 inhibitor
  • Inhibits USP7 with an EC50 of 8.01 μM
  • Inhibits USP47 with an EC50 of 8.74 μM
  • Shows antiproliferative activity against human HCT116 cells
  • Induces p53-mediated apoptosis in neuroblastoma cells
  • Exhibits antitumor effects in xenograft mouse models
Catalog No. 50-003-65540