Learn More
Abcam P 22077, 10MG
Supplier:  Abcam AB27338310MG
Encompass_Preferred
MW 315.3 Da, Purity >98%. Selective USP7 (IC₅₀ = 8 μM) inhibitor. Active against closely related USP47. Selective versus SENP2c0re (IC₅₀ >50 μM) and DENI (IC₅₀ >50 μM). Destabilizes Claspin. Inhibits neuroblastoma growth by inducing p53-mediated apoptosis. Inhibits Tip60 deubiquitination.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2960