Learn More
SPEX CERTIPREP LLC 3P P. aeruginosa High CFU Range (NCTC 12951), 20/pk
Supplier:  SPEX CERTIPREP LLC 3P 1066220X
Encompass_Preferred
(NSI by Zeptometrix) A lyophilized pure culture of P. aeruginosa preserved in our proprietary Microgel-Flash quick dissolve matrix. Formulated at 1000-2000cfu levels and supplied in packages of 10 or 20 individually vacuum sealed pellets. Designed for use as a negative control for water coliform analysis or as a positive quantitative control for water pseudomonas analysis.
Catalog No. NC1390522