Learn More
Sigma Aldrich Fine Chemicals Biosciences p-Aminobenzamidine-Agarose (1:1 suspension in a 1.0 M NaCl + 20% ethanol solution) | MFCD00165084 | 10ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A833210ML
Encompass
p-Aminobenzamidine-Agarose (1:1 suspension in a 1.0 M NaCl + 20% ethanol solution) | MFCD00165084 | 10ML
Catalog No. NC2338536