Learn More
Medchemexpress LLC p-Anisic acid | 100-09-4 | 152.15 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN13945G
Encompass_Preferred
p-Anisic acid (4-Methoxybenzoic acid) is an orally available tyrosinase inhibitor with antioxidant, anti-anxiety, anti-inflammatory, anti-tumor, anti-diabetic, and preservative properties. It can be utilized as a preservative in the cosmetics field.
  • Orally available tyrosinase inhibitor
  • Possesses antioxidant properties
  • Exhibits anti-anxiety effects
  • Anti-inflammatory activity
  • Demonstrates anti-tumor properties
  • Has anti-diabetic effects
  • Acts as a preservative for cosmetics
  • Stable under recommended storage conditions
  • Can be stored at room temperature for 3 years, or in solvent at -80°C for 2 years, -20°C for 1 year
Catalog No. 50-004-14735