Learn More
Medchemexpress LLC p-Anisic acid | 100-09-4 | 152.15 | 50 G
SDP
Supplier:  Medchemexpress LLC HYN139450G
Encompass_Preferred
p-Anisic acid (4-Methoxybenzoic acid) is an orally available tyrosinase inhibitor that possesses antioxidant, anti-anxiety, anti-inflammatory, anti-tumor, anti-diabetic, and preservative properties. It can be used as a preservative in the cosmetics field.
  • Orally available tyrosinase inhibitor.
  • Has antioxidant properties.
  • Exhibits anti-anxiety effects.
  • Possesses anti-inflammatory properties.
  • Demonstrates anti-tumor activity.
  • Shows anti-diabetic effects.
  • Can be used as a preservative in the cosmetics field.
Catalog No. 50-004-15760